MilliporeSigma
  • Anti-bilirubin monoclonal antibody. III. Preparation and properties of monoclonal antibodies to unconjugated bilirubin-IX alpha.
Anti-bilirubin monoclonal antibody. III. Preparation and properties of monoclonal antibodies to unconjugated bilirubin-IX alpha.
Biochimica et biophysica acta (1991-04-09)
Y Okamura, M Yamazaki, T Yamaguchi, Y Komoda, A Sugimoto, H Nakajima
ABSTRACT
Anti-bilirubin-IX alpha monoclonal antibodies exclusively specific for unconjugated bilirubin-IX alpha are prepared and characterized. Using modified MBS (metamaleimidobenzoyl-N-hydroxysuccinimide ester) method, bilirubin-IX alpha was covalently coupled to bovine serum albumin (BSA) retaining its intramolecular hydrogen bonds as well as three-dimensional structure. Somatic cell fusion was performed between murine spleen cells immunized with the bilirubin-IX alpha-BSA and murine myeloma P3-X63-Ag8-U1 cells. After screening assay, 58 clones were identified which were producing antibodies not to albumin nor MBS, but to haptenic bilirubin-IX alpha. In inhibition analysis, the antibodies in the culture supernatant reacted only with bilirubin-IX alpha to a varying degree, but did not react with conjugated bilirubin-IX alpha, including bilirubin-IX alpha monoglucuronide, bilirubin-IX alpha diglucuronide, and ditauronic bilirubin-IX alpha.
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Pricing and availability is not currently available.