MilliporeSigma
[Chemical constituents of roots of Boehmeria nivea].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-01-15)
Qiongming Xu, Guoqing Chen, Jinying Fan, Mengjia Zhang, Xia Li, Shilin Yang, Xiaoran Li
ABSTRACT
To investigate the chemical constituents from roots of Boehmeria nivea. The constituents were isolated by repeated column chromatography and preparative liquid chromatography; and their structures were elucidated by chemical properties and spectroscopic analyses. Seven compounds were isolated and their structures were identified as tormentic acid (1), hederagenin (2), maslinic acid (3), 2alpha-hydroxyursolic acid (4), trans-p-hydroxycinamic acid (5), 2,4,4'-trihydroxychalcone (6), rutin (7). Compounds 1-6 were obtained from this genus for the first time.
Sigma-Aldrich
Hederagenin, ≥97% (HPLC)