MilliporeSigma
  • Black tongue secondary to bismuth subsalicylate: case report and review of exogenous causes of macular lingual pigmentation.
Black tongue secondary to bismuth subsalicylate: case report and review of exogenous causes of macular lingual pigmentation.
Journal of drugs in dermatology : JDD (2009-12-24)
Philip R Cohen
ABSTRACT
Macular pigmentation of the tongue can be acquired following exposure to exogenous agents. Black lingual hyperpigmentation was observed during the full body skin examination of a man with a history of recurrent metastatic malignant melanoma. His tongue spontaneously returned to its normal pink color later that day. Bismuth subsalicylate (Pepto-Bismol) was suspected as the pigment-inducing agent; subsequently, re-challange with the antacid confirmed it to be the cause of his acquired, albeit transient, black tongue. The ingestion of medications, including other antacids, analgesics, antidepressants, antihypertensives and several antimicrobials has been associated with the development of acquired macular lingual pigmentation. In addition, hyperpigmentation of the tongue has been observed following the deposition of amalgam and the injection of local anesthesia or doxorubicin or interferon alpha and ribavirin. Also, inhalation of heroin and methaqualone vapors or tobacco has resulted in lingual hyperpigmentation. All of the patients with acquired macular lingual hyperpigmentation had tongues with a smooth surface without enlargement of the filiform papillae. Many of the individuals with hyperpigmented tongue had either black or dark skin color. The onset of tongue pigmentation varied from less than one day to several years after initial exposure to the associated exogenous agent. The color of the tongue usually returned to normal after the pigment-inducing agent was discontinued.
Sigma-Aldrich
Bismuth(III) subsalicylate, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 150x150mm, thickness 0.05mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Pricing and availability is not currently available.
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Pricing and availability is not currently available.