MilliporeSigma
Asymmetric synthesis of alpha-tocopherol.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-12-17)
Urs Hengartner, Antoinette Chougnet, Kegang Liu, Wolf-D Woggon
ABSTRACT
Alpha-tocopherol was synthesized from a chiral intermediate alpha-hydroxy ester by means of two ring-closing methods to yield the chromanol in 94% diastereomeric excess.
Sigma-Aldrich
2,2,5,7,8-Pentamethyl-6-chromanol, 97%
Pricing and availability is not currently available.