MilliporeSigma
  • Antifouling surface layers for improved signal-to-noise of particle-based immunoassays.
Antifouling surface layers for improved signal-to-noise of particle-based immunoassays.
Langmuir : the ACS journal of surfaces and colloids (2009-11-26)
Annie Chen, Darby Kozak, Bronwyn J Battersby, Robin M Forrest, Nathalie Scholler, Nicole Urban, Matt Trau
ABSTRACT
A 10-fold improvement in the signal-to-noise (S/N) ratio of an optically encoded silica particle-based immunoassay was achieved through incorporating a protein resistant poly(ethylene glycol) (PEG) surface layer and optimizing antibody immobilization conditions. PEG was activated using 2,2,2-trifluoroethanesulfonyl chloride (tresyl) and required a minimum reaction time of 1.5 h. The activated PEG had a reactive half-life of approximately 5 h when stored in acidified dimethyl sulfoxide (DMSO). By increasing the protein incubation time and concentration, a maximum antibody loading on the particle surface of 1.6 x 10(-2) molecules per nm(2) was achieved. The assay S/N ratio was assessed using a multiplexed multicomponent optically encoded species-specific immunoassay. Encoded particles were covalently grafted or nonspecifically coated with either bovine or mouse IgG for the simultaneous detection of complementary anti-IgG "target" or uncomplementary anti-IgG "noise". The versatility and potential as a serum-based assay platform was demonstrated by immobilizing either a polyclonal antibody or an engineered single-chain variable fragment (scFv) capture probe on particles for the detection of the ovarian cancer biomarker, mesothelin (MSLN). The MLSN antigen was spiked into PBS buffer or 50% human serum. Both capture probe orientations, and media conditions showed similar low level detection limits of 5 ng/mL; however, a 40% decrease in maximum signal intensity was observed for assays run in 50% serum.
Sigma-Aldrich
2,2,2-Trifluoroethanesulfonyl chloride, 99%