MilliporeSigma
  • GC separation of amino acid enantiomers via derivatization with heptafluorobutyl chloroformate and Chirasil-L-Val column.
GC separation of amino acid enantiomers via derivatization with heptafluorobutyl chloroformate and Chirasil-L-Val column.
Journal of separation science (2009-10-21)
Helena Zahradnícková, Petr Husek, Petr Simek
ABSTRACT
Heptafluorobutyl chloroformate (HFBCF), a recently introduced derivatization reagent, was examined in enantioseparation of amino acids (AAs) by GC. Twenty proteinogenic AAs, plus ornithine, cystine and 4-fluorophenylalanine (internal standard) were treated with the reagent and separation properties of the derivatives were assessed on a Chirasil-Val capillary column. Nineteen AA enantiomers were efficiently separated in 43 min except proline, arginine and cystine. The HFBCF derivatives of the studied DL-AAs show improved separation over other chloroformate-based derivatives hitherto reported. A combination of the improved and faster separation with a simple derivatization protocol, involving an immediate one-step reaction-extraction in two-phase aqueous-organic medium, and low elution temperatures extend application of HFBCF to chiral AA analysis.
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Supelco
Heptafluorobutyric acid, suitable for ion chromatography, ≥99.5% (GC)
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography