Merck
  • Robust recognition of malonate and 2-amino-4-picolinium in conjunction with M(II) as a triad (M = Ni/Co/Mn): role of this highly stable hydrogen-bonded motif in driving supramolecular self-assembly.
Robust recognition of malonate and 2-amino-4-picolinium in conjunction with M(II) as a triad (M = Ni/Co/Mn): role of this highly stable hydrogen-bonded motif in driving supramolecular self-assembly.
Dalton transactions (Cambridge, England : 2003) (2009-09-18)
Somnath Ray Choudhury, Biswajit Dey, Suranjana Das, Arturo Robertazzi, Atish Dipankar Jana, Chih-Yuan Chen, Hon Man Lee, Patrick Gamez, Subrata Mukhopadhyay
ABSTRACT
The crystal lattice of the three isostructural compounds , (C(6)H(8)N(2)H)(2)[M(C(3)H(2)O(4))(2)(H(2)O)(2)].4H(2)O (C(6)H(8)N(2)H = protonated 2-amino-4-picoline, M = Ni/Co/Mn, C(3)H(4)O(4) = malonate dianion; hereafter, malonate) is formed by supramolecular 2D layers. Hydrogen-bonding, pi...pi and lone pair...pi interactions play crucial role in organizing monomeric [M(II)(mal)(2)(H(2)O)(2)] units into 2D sheets along the ab plane, through the self-association between two different supramolecular building blocks, namely a tetrameric water cluster including metal-coordinated water molecules, and R(2)(2)(8) and R(2)(2)(7) hydrogen-bonded recognition synthons between 2-amino-4-picolinium and malonate. DFT calculations clearly show that the robust 2-amino-4-picolinium/malonate hydrogen-bonded motif drives the self-assembly of the supramolecular network observed.
Sigma-Aldrich
2-Amino-4-methylpyridine, 99%