MilliporeSigma
  • A 2-aminobenzimidazole that inhibits and disperses gram-positive biofilms through a zinc-dependent mechanism.
A 2-aminobenzimidazole that inhibits and disperses gram-positive biofilms through a zinc-dependent mechanism.
Journal of the American Chemical Society (2009-07-23)
Steven A Rogers, Robert W Huigens, Christian Melander
ABSTRACT
A number of 2-aminobenzimidazole derivatives were synthesized and screened for their ability to inhibit and disperse bacterial biofilms. From these compounds, a lead 2-aminobenzimidazole was identified that both inhibited and dispersed MRSA, vancomycin-resistant Enterococcus faecium, and Staphylococcus epidermidis biofilms. Mechanistic studies showed that the activity is Zn(II)-dependent, potentially via a direct zinc-chelating mechanism.
Sigma-Aldrich
2-Aminobenzimidazole, 97%
Pricing and availability is not currently available.