- Synthesis of the tetracyclic core of the neomangicols using a late-stage indene alkylation.
Synthesis of the tetracyclic core of the neomangicols using a late-stage indene alkylation.
Organic letters (2009-06-17)
Jessica L Wood, Brian G Pujanauski, Richmond Sarpong
PMID19527042
ABSTRACT
A general approach to the tetracyclic core of the neomangicol natural products via a late-stage indene alkylation reaction is presented. This strategy sets the stage for access to the neomangicol family and, in addition, provides a potential biogenetically inspired entry to the mangicol natural products.
Product Description
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
Indene, contains 50-100 ppm tert-butylcatechol as stabilizer, technical grade, ≥90%
Pricing and availability is not currently available.