MilliporeSigma
  • Experimental stroke protection induced by 4-hydroxybenzyl alcohol is cancelled by bacitracin.
Experimental stroke protection induced by 4-hydroxybenzyl alcohol is cancelled by bacitracin.
Neuroscience research (2009-05-12)
Elodie Descamps, Maud Petrault-Laprais, Pierre Maurois, Nicole Pages, Pierre Bac, Régis Bordet, Joseph Vamecq
ABSTRACT
Induction of protein disulfide isomerase (PDI) is validated as a main mechanism by which 4-hydroxybenzyl alcohol (4-HBA), an active principle of Gastrodia elata Blume, reduces cerebral infarct volumes in a murine model of focal brain ischemia/reperfusion. In contrast to its position isomers, i.e. 3-hydroxybenzyl alcohol (3-HBA) and 2-hydroxybenzyl alcohol (2-HBA), and to aliphatic diols (1,4-butanediol and 1,5-pentanediol), 4-HBA administered intravenously at 25 mg/kg protected mice, significantly reducing total, cortical and sub-cortical infarct volumes by 42, 28 and 55%, respectively. All compounds, 4-HBA included, were devoid of antioedematous properties. Only the stroke protective 4-HBA, but neither 3-HBA nor 2-HBA, was capable of significantly inducing PDI in intact mouse brains. Stroke protection was fully prevented by bacitracin (500 mg/kg), a known inhibitor of PDI, which, without affecting basal brain PDI levels, altered the ability of 4-HBA to induce significantly PDI in intact brains. Taken as a whole, our data indicate that stroke protection induced by 4-HBA involves PDI as a key player, making this protein a valuable target to control brain injury disorders. The fact that 4-HBA, at doses up to 200mg/kg, was devoid of neurotoxicity in the rotarod test is also a decisive element to promote the neuroprotective use of this plant compound.
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG
Sigma-Aldrich
4-Hydroxybenzyl alcohol, 99%