MilliporeSigma
  • Synthesis and biological evaluation of penem inhibitors of bacterial signal peptidase.
Synthesis and biological evaluation of penem inhibitors of bacterial signal peptidase.
Bioorganic & medicinal chemistry letters (2009-05-12)
David A Harris, Michael E Powers, Floyd E Romesberg
ABSTRACT
We report the first synthesis of a 5S penem, known to bind bacterial type I signal peptidase, from the commercially available and inexpensive 6-aminopenicillanic acid. We report the first in vivo activity of the compound and use structure-activity relationship studies to begin to define the determinants of signal peptidase binding and also to begin to optimize the penem as an antibiotic.
Sigma-Aldrich
(+)-6-Aminopenicillanic acid, 96%