MilliporeSigma
  • Arsenic speciation in natural and contaminated waters using CZE with in situ derivatization by molybdate and direct UV-detection.
Arsenic speciation in natural and contaminated waters using CZE with in situ derivatization by molybdate and direct UV-detection.
Electrophoresis (2009-03-31)
Olga S Koshcheeva, Olga V Shuvaeva, Lidiya I Kuznetzova
ABSTRACT
A simple and sensitive procedure for simultaneous determination of arsenate, arsenite, monomethylarsonate and dimethylarsinate (DMA) ions in waters using CZE with chemical derivatization in situ and UV-detection at 250 nm was developed. The separation was performed in a fused-silica capillary using solution containing sodium molybdate and sodium perchlorate as electrolyte. Molybdate forms heteropolycomplexes with arsenic species in low acidic media, while sodium perchlorate masks silicate ion. The analysis conditions were optimized; the best results were achieved with the electrolyte consisting of 10 mM Na(2)MoO(4) and 10 mM NaClO(4) at pH 3.0 using negative voltage and pneumatic injection of the sample. Nevertheless, the signal of arsenite ion was not detected, probably because of its instability. Arsenite ion was quantified as a difference between arsenate ion contents after and before oxidation by bromine water. The detection limits for the fresh water at the level of 5.0 microg/L for As(III) and As(V), 16 microg/L for DMA and 20 microg/L for MMA were achieved. The reproducibility varied in the range of 0.06-0.25 relative units. To reduce the interferences of the sample salinity an addition of organic substances and isotachophoretic effect were used.
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis