MilliporeSigma
  • Novel use of a whole cell E. coli bioreporter as a urinary exposure biomarker.
Novel use of a whole cell E. coli bioreporter as a urinary exposure biomarker.
Environmental science & technology (2009-02-26)
Ceri Lewis, Siham Beggah, Chris Pook, Carlos Guitart, Clare Redshaw, Jan Roelof van der Meer, James W Readman, Tamara Galloway
ABSTRACT
Bacterial bioreporters have substantial potential for contaminant assessment but their real world application is currently impaired by a lack of sensitivity. Here, we exploit the bioconcentration of chemicals in the urine of animals to facilitate pollutant detection. The shore crab Carcinus maenas was exposed to the organic contaminant 2-hydroxybiphenyl, and urine was screened using an Escherichia coli-based luciferase gene (luxAB) reporter assay specific to this compound. Bioassay measurements differentiated between the original contaminant and its metabolites, quantifying bioconcentration factors of up to one hundred-fold in crab urine. Our results reveal the substantial potential of using bacterial bioreporter assays in real-time monitoring of biological matricesto determine exposure histories, with wide ranging potential for the in situ measurement of xenobiotics in risk assessments and epidemiology.
Sigma-Aldrich
2-Phenylphenol, 99%
Supelco
2-Phenylphenol, PESTANAL®, analytical standard