MilliporeSigma
  • Facilitated transport of Cr(III) through activated composite membrane containing di-(2-ethylhexyl)phosphoric acid (DEHPA) as carrier agent.
Facilitated transport of Cr(III) through activated composite membrane containing di-(2-ethylhexyl)phosphoric acid (DEHPA) as carrier agent.
Journal of hazardous materials (2008-11-28)
Gulsin Arslan, Ali Tor, Yunus Cengeloglu, Mustafa Ersoz
ABSTRACT
The facilitated transport of chromium(III) through activated composite membrane (ACM) containing di-(2-ethylhexyl) phosphoric acid (DEHPA) was investigated. DEHPA was immobilised by interfacial polymerisation on polysulfone layer which was deposited on non-woven fabric by using spin coater. Then, ACM was characterised by using scanning electron microscopy (SEM), contact angle measurements and atomic force microscopy (AFM). Initially, batch experiments of liquid-liquid distribution of Cr(III) and the extractant (DEHPA) were carried out to determine the appropriate pH of the feed phase and the results showed that maximum extraction of Cr(III) was achieved at a pH of 4. It was also found that Cr(III) and DEHPA reacted in 1/1 molar ratio. The effects of Cr(III) (in feed phase), HCl (in stripping phase) and DEHPA (in ACM) concentrations were investigated. DEHPA concentration varies from 0.1 to 1.0M and it was determined that the transport of Cr(III) increased with the carrier concentration up to 0.8M. It was also observed that the transport of Cr(III) through the ACM tended to increase with Cr(III) and HCl concentrations. The stability of ACM was also confirmed with replicate experiments.
Sigma-Aldrich
Bis(2-ethylhexyl) phosphate, 97%