MilliporeSigma
  • In vitro screening for aryl hydrocarbon receptor agonistic activity in 200 pesticides using a highly sensitive reporter cell line, DR-EcoScreen cells, and in vivo mouse liver cytochrome P450-1A induction by propanil, diuron and linuron.
In vitro screening for aryl hydrocarbon receptor agonistic activity in 200 pesticides using a highly sensitive reporter cell line, DR-EcoScreen cells, and in vivo mouse liver cytochrome P450-1A induction by propanil, diuron and linuron.
Chemosphere (2008-10-07)
Shinji Takeuchi, Mitsuru Iida, Hisatoshi Yabushita, Tadashi Matsuda, Hiroyuki Kojima
ABSTRACT
The aryl hydrocarbon receptor (AhR) is a ligand-dependent transcription factor that regulates genes involved in xenobiotic metabolism, cellular proliferation and differentiation. In this study, we have developed a highly sensitive AhR-mediated reporter cell line, DR-EcoScreen cells, which are mouse hepatoma Hepa1c1c7 cells stably transfected with a reporter plasmid containing seven copies of dioxin-responsive element. Using these DR-EcoScreen cells, we performed the reporter gene assay and characterized the AhR agonistic activities of 200 pesticides (29 organochlorines, 11 diphenyl ethers, 56 organophosphorus pesticides, 12 pyrethroids, 22 carbamates, 12 acid amides, 7 triazines, 6 ureas, and 45 others). Eleven of the 200 pesticides (acifluorfen-methyl, bifenox, chlorpyrifos, isoxathion, quinalphos, chlorpropham, diethofencarb, propanil, diuron, linuron, and prochloraz) showed AhR-mediated transcriptional activity. In particular, three herbicides (propanil, diuron, and linuron) have a common chemical structure and showed more potent agonistic activity than other pesticides. To investigate the in vivo effects, we examined the gene expression of AhR-inducible cytochrome P450 1As (CYP1As) in the liver of female C57BL/6 mice intraperitoneally injected with these three herbicides (300 mg kg(-1)) by quantitative RT-PCR, resulting in induction of significant high levels of CYP1A1 and CYP1A2 mRNAs. This indicates that propanil, diuron and linuron possess AhR-mediated transactivation effect in vivo as well as in vitro. Through the present study, we demonstrated that DR-EcoScreen cells are useful for sensitive, rapid and simple identification of AhR agonists among a large number of environmental chemicals.
Product Description
Supelco
Propanil, PESTANAL®, analytical standard
Pricing and availability is not currently available.