MilliporeSigma
  • Fast, noninvasive and simultaneous near-infrared spectroscopic characterisation of physicochemical stationary phases' properties: from silica particles towards monoliths.
Fast, noninvasive and simultaneous near-infrared spectroscopic characterisation of physicochemical stationary phases' properties: from silica particles towards monoliths.
Journal of separation science (2008-08-12)
Christine H Petter, Nico Heigl, Günther K Bonn, Christian W Huck
ABSTRACT
The design of novel stationary phases is a permanent demanding challenge in chromatographic separation science to enable analysis with enhanced selectivity, specificity and speed. Therefore, the characterisation of chemical and physical properties is next to calculation of chromatographic parameters essential. Conventionally, chemical parameters including surface coverage are determined by burning combustion or frontal analysis, physical parameters including particle size, pore size, pore volume and surface area are determined by SEM, mercury intrusion porosimetry (MIP) and Brunauer-Emmett-Teller (BET). All these methods are time consuming, invasive and require besides special equipment some special trained laboratory staff. Therefore, we introduced near-infrared spectroscopy (NIRS) as a noninvasive, easy-to-handle technology with wavenumber ranging from 4000 to 10,000 cm(-1) enabling analysis within only a few seconds at higher precision than the conventional methods. Investigated materials comprise porous and nonporous silica gel, carbon-based nanomaterials (fullerenes), polymer beads and monoliths. Different carriers themselves and their kind of derivatisations (RP, normal-phase, ion-exchanger, IMAC (immobilised metal affinity chromatography), affinity) can be determined by applying principal component analysis (PCA) of recorded spectra. Partial least square regression (PLSR) enables the determination of particle size, pore size, pore volume, porosity, total porosity and surface area with one single measurement. For the optimised design of well-defined polymer beads and monoliths, real-time in situ monitoring to control, e. g. particle and pore sizes as well as monomer content during the polymerisation process, can be extremely helpful. In this article, the advantages of this fast, noninvasive high-throughput NIRS methods are summarised, discussed in detail and different applications of the individual characterised materials are shown.
Sigma-Aldrich
Methylstyrene, 60% meta, 40% para and 1% ortho, 99%, contains ~50 ppm 4-tert-butylcatechol as inhibitor