MilliporeSigma
  • Protective effects of protopine on hydrogen peroxide-induced oxidative injury of PC12 cells via Ca(2+) antagonism and antioxidant mechanisms.
Protective effects of protopine on hydrogen peroxide-induced oxidative injury of PC12 cells via Ca(2+) antagonism and antioxidant mechanisms.
European journal of pharmacology (2008-07-08)
Xianghua Xiao, Juntian Liu, Jingwen Hu, Xiuping Zhu, Hua Yang, Chaoyun Wang, Yuanhui Zhang
ABSTRACT
Calcium and lipid peroxidation play important roles in oxidative stress-induced cellular injury and apoptosis, which ultimately cause cell death. In this study we examined whether protopine had a neuroprotection against H(2)O(2)-induced injury in PC12 cells. Pretreatment of PC12 cells with protopine improved the cell viability, enhanced activities of superoxide dismutase, glutathione peroxidase and catalase, and decreased malondialdehyde level in the H(2)O(2) injured cells. Protopine also reversed the increased intracellular Ca(2+) concentration and the reduced mitochondrial membrane potential caused by H(2)O(2) in the cells. Furthermore, protopine was able to inhibit caspase-3 expression and cell apoptosis induced by H(2)O(2). In summary, this study demonstrates that protopine is able to relieve H(2)O(2)-induced oxidative stress and apoptosis in PC12 cells, at least in part, by Ca(2+) antagonism and antioxidant mechanisms.
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid