MilliporeSigma
  • Cardioprotection with alpha-tocopheryl phosphate: amelioration of myocardial ischemia reperfusion injury is linked with its ability to generate a survival signal through Akt activation.
Cardioprotection with alpha-tocopheryl phosphate: amelioration of myocardial ischemia reperfusion injury is linked with its ability to generate a survival signal through Akt activation.
Biochimica et biophysica acta (2008-06-17)
Subhendu Mukherjee, Istvan Lekli, Manika Das, Angelo Azzi, Dipak K Das
ABSTRACT
The emerging potential of alpha-tocopheryl phosphate, a phosphoric acid ester of alpha-tocopherol, in health benefits was tested gavaging this compound (5 mg/kg body wt) to a group of rats for a period of thirty days while the control rats were given water only. After thirty days, the rats were sacrificed, the hearts excised, and the isolated hearts were perfused by working mode. Both control and experimental hearts were subjected to 30-min global ischemia followed by 2 h of reperfusion. The tocopheryl phosphate fed rats exhibited significant cardioprotection as evidenced by improved ventricular performance and reduced myocardial infarct size and cardiomyocyte apoptosis. Supplementation with alpha-tocopheryl phosphate converted MAP kinase-induced death signal into a survival signal by enhancing anti-apoptotic p42/44 ERK kinase and p38 MAPKbeta and reducing pro-apoptotic proteins p38 MAPKalpha and JNK. In concert, the phosphorylation of pro-apoptotic c-Src was also reduced. Tocopheryl phosphate increased the DNA binding of the redox-sensitive transcription factor NFkappaB and potentiated the activation of anti-death protein Bcl-2 and survival signaling protein Akt. The results of this study demonstrated for the first time that tocopheryl phosphate could ameliorate myocardial ischemic reperfusion injury by converting ischemia/reperfusion-mediated death signal into a survival signal by modulating MAP kinase signaling.
Sigma-Aldrich
(±)-α-Tocopherol phosphate disodium salt, water-soluble α-tocopherol analog
Pricing and availability is not currently available.