MilliporeSigma
  • Microbial hydroxylation of o-bromophenylacetic acid: synthesis of 4-substituted-2,3-dihydrobenzofurans.
Microbial hydroxylation of o-bromophenylacetic acid: synthesis of 4-substituted-2,3-dihydrobenzofurans.
Journal of industrial microbiology & biotechnology (2008-05-23)
Prashant P Deshpande, Venkata B Nanduri, Annie Pullockaran, Hamish Christie, Richard H Mueller, Ramesh N Patel
ABSTRACT
Microbial hydroxylation of o-bromophenylacetic acid provided 2-bromo-5-hydroxyphenylacetic acid. This enabled a route to the key intermediate 4-bromo-2,3-dihydrobenzofuran for synthesizing a melatonin receptor agonist and sodium hydrogen exchange compounds. Pd-mediated coupling reactions of 4-bromo-2,3-dihydrobenzofuran provided easy access to the 4-substituted-2,3-dihydrobenzofurans.
Sigma-Aldrich
2,3-Dihydrobenzofuran, 99%