MilliporeSigma
  • Insensitivity of NMRI mice to selective serotonin reuptake inhibitors in the tail suspension test can be reversed by co-treatment with 5-hydroxytryptophan.
Insensitivity of NMRI mice to selective serotonin reuptake inhibitors in the tail suspension test can be reversed by co-treatment with 5-hydroxytryptophan.
Psychopharmacology (2008-05-23)
Jacob P R Jacobsen, Elsebet Ø Nielsen, Rene Hummel, John Paul Redrobe, Naheed Mirza, Pia Weikop
ABSTRACT
Exploring differences between mouse strains in drug effects in models of antidepressant-like activity may provide clues to the neurobiology of antidepressant responses. The objective of this study was to explore whether insensitivity to selective serotonin reuptake inhibitors (SSRIs) in NMRI mice in the tail suspension test can be related to 5-hydroxytryptamine (5-HT) function. We compared NMRI and C57Bl/6 mice, a SSRI-sensitive strain, in the tail suspension test following citalopram, paroxetine, or fluoxetine and determined 5-HT transporter (5-HTT) densities, 5-HT tissue and extracellular levels, 5-HT synthesis, tryptophan hydroxylase 2 (TPH2) genotypes and hypothermia induced by the 5-HT(1A) agonist 8-OH-DPAT. In NMRI mice, we tested if co-treatment with 5-HTP would increase 5-HT levels and confer SSRI sensitivity in the tail suspension test. C57Bl/6, but not NMRI, mice responded to SSRIs in the tail suspension test. 5-HTT densities in the frontal cortex and hippocampus were similar between the strains. NMRI mice had lower tissue 5-HT levels in these regions and decreased extracellular 5-HT in the frontal cortex at baseline and following citalopram. C57Bl/6 mice were more sensitive to 8-OH-DPAT-induced hypothermia. Both strains had the 1473C TPH2 genotype and similar 5-HT synthesis. In NMRI mice, 5-HTP co-treatment restored the tail suspension and extracellular 5-HT responses to SSRIs to levels equivalent to those seen in C57Bl/6 mice. Low 5-HT function in NMRI mice may account for their insensitivity to SSRIs in the tail suspension test. As the tail suspension test is a predictor of clinical efficacy, the current data suggest that 5-HTP adjunct treatment may benefit SSRI treatment refractory patients.
Sigma-Aldrich
Nisoxetine hydrochloride, solid, ≥98% (HPLC)