MilliporeSigma
  • Ground state spin-switching via targeted structural distortion: twisted single-molecule magnets from derivatised salicylaldoximes.
Ground state spin-switching via targeted structural distortion: twisted single-molecule magnets from derivatised salicylaldoximes.
Dalton transactions (Cambridge, England : 2003) (2008-03-29)
Constantinos J Milios, Stergios Piligkos, Euan K Brechin
ABSTRACT
The use of derivatised salicylaldoximes in Mn chemistry has led to the isolation of a plethora of beautiful new SMMs ranging in nuclearity from three to eight and with spin ground states as large as S = 12-including a Mn6 complex with the largest energy barrier to magnetisation reversal yet reported. The deliberate chemically-induced structural distortion of the [Mn6] molecule allows the isolation of analogous family members displaying remarkably different magnetic properties and this in turn allows for a rare semi-quantitative magneto-structural correlation which enables prediction of the magnetic properties of new family members.
Sigma-Aldrich
Salicylaldoxime, ≥98.0% (NT)