MilliporeSigma
Shiftless nuclear magnetic resonance spectroscopy.
The Journal of chemical physics (2008-02-13)
Chin H Wu, Stanley J Opella
ABSTRACT
The acquisition and analysis of high resolution one- and two-dimensional solid-state nuclear magnetic resonance (NMR) spectra without chemical shift frequencies are described. Many variations of shiftless NMR spectroscopy are feasible. A two-dimensional experiment that correlates the dipole-dipole and dipole-dipole couplings in the model peptide , (15)N labeled N-acetyl-leucine is demonstrated. In addition to the resolution of resonances from individual sites in a single crystal sample, the bond lengths and angles are characterized by the two-dimensional powder pattern obtained from a polycrystalline sample.
Sigma-Aldrich
N-Acetyl-L-leucine, ReagentPlus®, 99%