MilliporeSigma
  • Stereoselective quaternization of alpha-amino phenylacetonitriles mediated by a remote sulfinyl group.
Stereoselective quaternization of alpha-amino phenylacetonitriles mediated by a remote sulfinyl group.
The Journal of organic chemistry (2007-07-17)
José Luis García Ruano, Ana M Martín-Castro, Francisco Tato, Inés Alonso
ABSTRACT
Enantiomerically pure alpha-substituted alpha-amino phenylacetonitriles have been readily prepared from 2-p-tolylsulfinylbenzaldimines following a two-step sequence: a moderately stereoselective hydrocyanation of the imines and a completely stereoselective quaternization of the resulting diastereoisomeric mixture of alpha-amino phenylacetonitriles with different alkylating or acylating reagents in the presence of KHMDS. Theoretical calculations support a stereoselectivity control exerted by the remote sulfinyl group, as long as it is responsible for the conformational preferences of the benzyllithium intermediates, which suffer the attack of the electrophiles to the less hindered diastereotopic face.
Sigma-Aldrich
Benzyl cyanide, 98%