MilliporeSigma
  • Simultaneous analysis of D- and L-serine in cerebrospinal fluid by use of HPLC.
Simultaneous analysis of D- and L-serine in cerebrospinal fluid by use of HPLC.
Clinical chemistry (2007-06-26)
Rama Sethuraman, Malathi G Krishnamoorthy, Tat-Leang Lee, Eugene Hern C Liu, Siau Chiang, Wataru Nishimura, Masato Sakai, Toshiaki Minami, Shinro Tachibana
ABSTRACT
D-Serine is a coagonist for the glycine-binding site of the N-methyl-D-aspartate receptors and has been implicated in various neuropsychiatric functions such as learning, memory, and nociception, as well as schizophrenia and Alzheimer disease. We developed an HPLC method for D- and L-serine in cerebrospinal fluid (CSF). The dabsylated racemic serine peak, automatically collected using a previously reported HPLC separation process for CSF amino acids, was desalted and subjected to a chiral resolution HPLC step with a Sumichiral column using an ultraviolet-visible detector. The limits of quantification (signal-to-noise ratio = 10) for D- and L-serine were 0.8 and 1.3 micromol/L, respectively. The mean imprecision values (CVs) for within-day measurements of D- and L-serine were 2.1% and 1.8%, respectively, and for between-day were 6.2% and 6.6%. Mean recovery of CSF serine (sum of D-serine + L-serine) applied to the Sumichiral column was 87%. The mean (SD) d-serine concentrations in 45 CSF samples obtained from 16 patients with chronic pain due to degenerative osteoarthritis of the knees, 16 with postherpetic neuralgia, and 13 with no pain were, respectively, 3.97 (0.44), 1.85 (0.21), and 2.72 (0.32) micromol/L. D- and L-serine can be quantified with ultraviolet-visible detection of dabsyl derivatives. The dabsyl derivatives are stable and allow duplicate analysis of CSF samples in multisample runs.
Sigma-Aldrich
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride, ≥97.5% (AT)