MilliporeSigma
[Studies on the chemical constituents from Inula cappa (II)].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2007-06-02)
Qi-lei Guo, Jun-shan Yang, Jian-xun Liu
ABSTRACT
To study the chemical constituents of the root of Inula cappa. The compounds were isolated and purified by recrystallization and chromatography with silica gel column and Sephadex LH-20 column. Their structures were identified by physico-chemical properties and spectral analysis. Seven compounds were isolated as scopolin (I), octacosanoic acid (II), tritriacontane (III), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxydocosanosylamino]-octadecane-1,3,4-triol(IV),(2S,3S,4R,8E)-2-[(2'R)-2'-hydroxytricosanosylamino]-octadecane-1,3,4-triol(V), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxytetracosanosylamino]-octadecane-1,3,4-triol(VI), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxypentacosanosylamino]-octadecane-1,3,4-triol(VIII), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxyhexacosanosylamino]-octadecane-1,3,4-triol(VIII). All compounds were isolated from Inula cappa for the first time.
Sigma-Aldrich
Octacosanoic acid, synthetic, ≥98%