MilliporeSigma
  • Discovery of para-alkylthiophenoxyacetic acids as a novel series of potent and selective PPARdelta agonists.
Discovery of para-alkylthiophenoxyacetic acids as a novel series of potent and selective PPARdelta agonists.
Bioorganic & medicinal chemistry letters (2007-05-26)
Rui Zhang, Aihua Wang, Alan DeAngelis, Patricia Pelton, Jun Xu, Peifang Zhu, Lubing Zhou, Keith Demarest, William V Murray, Gee-Hong Kuo
ABSTRACT
A novel series of potent and selective PPARdelta agonists, para-alkylthiophenoxyacetic acids, was identified. The synthesis and structure-activity relationships are described.
Sigma-Aldrich
(Phenylthio)acetic acid, 96%