MilliporeSigma
  • The effect of physico-chemical parameters and chemical compounds on the activity of beta-d-galactosidase (B-GAL), a marker enzyme for indicator microorganisms in water.
The effect of physico-chemical parameters and chemical compounds on the activity of beta-d-galactosidase (B-GAL), a marker enzyme for indicator microorganisms in water.
Chemosphere (2007-04-17)
V C Wutor, C A Togo, B I Pletschke
ABSTRACT
The presence of coliforms in polluted water was determined enzymatically (in situ) by directly monitoring the activity of beta-d-galactosidase (B-GAL) through the hydrolysis of the yellow chromogenic subtrate, chlorophenol red beta-d-galactopyranoside (CPRG), which produced a red chlorophenol red (CPR) product. The objectives of this study were to monitor the effect of compounds commonly found in the environment and used in water treatment on a B-GAL CPRG assay and to investigate the differences between the environmental B-GAL enzyme and the pure commercial enzyme. Environmental B-GAL was optimally active at pH 7.8. Two temperature optima were observed at 35 and 55 degrees C, respectively. B-GAL activity was strongly inhibited by silver and copper ions. While calcium and ferrous ions at lower concentrations (50-100mgl(-1)) increased the enzyme activity, a reduction was observed at higher concentrations (200mgl(-1)). Sodium hypochlorite, normally used in rural areas to disinfect water gradually decreased B-GAL activity at concentrations between 0 and 5600ppm for both the commercial and environmental enzymes. B-GAL from the environment behaved differently from its commercially available counterpart.
Sigma-Aldrich
Chlorophenol Red-β-D-galactopyranoside, ≥90% (HPLC)
Pricing and availability is not currently available.