MilliporeSigma
  • Characterization of cholinesterases in marbled sole, Limanda yokohamae, and their inhibition in vitro by the fungicide iprobenfos.
Characterization of cholinesterases in marbled sole, Limanda yokohamae, and their inhibition in vitro by the fungicide iprobenfos.
Marine environmental research (2007-02-16)
Jee-Hyun Jung, R F Addison, Won Joon Shim
ABSTRACT
Cholinesterases (ChEs) have been characterized in marbled sole (Limanda yokohamae) for use as a possible biomarker of pollution exposure. In brain, ChEs existed almost exclusively (>95%) as acetylcholinesterase (AChE) whereas in muscle, about 20-30% of ChE activity was in the form of butyrylcholinesterase (BChE; pseudocholinesterase). Acetylthiocholine and butyrylthiocholine (identified in mammalian studies as diagnostic substrates for AChE and BChE respectively) were hydrolyzed mainly, but not exclusively, by these enzymes. The inhibitors BW284C51 and iso-OMPA (identified in mammalian studies as diagnostic inhibitors of AChE and BChE respectively) were not specific for these enzymes in marbled sole. Brain AChE and muscle AChE and BChE were characterized in terms of their kinetic properties (KM etc.) and optimal conditions (substrate concentration, protein concentration, pH etc.) were established to allow routine assays of ChE activity to proceed under pseudo-first order conditions. The sensitivity of ChEs to a locally significant pesticide, iprobenfos (IBP; kitazin) was established in terms of IC50 concentrations. Brain AChE was relatively insensitive to IBP, but muscle AChE and BChE were sensitive to IBP concentrations in the high nM range. However, ambient IBP concentrations in Korean coastal waters are usually not high enough to cause detectable ChE inhibition in this species.
Product Description
Supelco
Iprobenfos, PESTANAL®, analytical standard
Pricing and availability is not currently available.