MilliporeSigma
  • Dissociation channels of the 1-buten-2-yl radical and its photolytic precursor 2-bromo-1-butene.
Dissociation channels of the 1-buten-2-yl radical and its photolytic precursor 2-bromo-1-butene.
The journal of physical chemistry. A (2006-07-13)
Johanna L Miller, Maria J Krisch, Laurie J Butler, Jinian Shu
ABSTRACT
The work presented here is the first in a series of studies that use a molecular beam scattering technique to investigate the unimolecular reaction dynamics of C(4)H(7) radical isomers. Photodissociation of the halogenated precursor 2-bromo-1-butene at 193 nm under collisionless conditions produced 1-buten-2-yl radicals with a range of internal energies spanning the predicted barriers to the unimolecular reaction channels of the radical. Resolving the velocities of the stable C(4)H(7) radicals, as well as those of the products, allows for the identification of the energetic onset of each dissociation channel. The data show that radicals with at least 30.7 +/- 2 kcal/mol of internal energy underwent C-C fission to form allene + methyl, and radicals with at least 36.7 +/- 4 kcal/mol of internal energy underwent C-H fission to form H + 1-butyne and H + 1,2-butadiene; both of these observed barriers agree well with the G3//B3LYP calculations of Miller. HBr elimination from the parent molecule was observed, producing vibrationally excited 1-butyne and 1,2-butadiene. In the subsequent dissociation of these C(4)H(6) isomers, the major channel was C-C fission to form propargyl + methyl, and there is also evidence of at least one of the possible H + C(4)H(5) channels. A minor C-Br fission channel produces 1-buten-2-yl radicals in an excited electronic state and with low kinetic energy; these radicals exhibit markedly different dissociation dynamics than do the radicals produced in their ground electronic state.
Sigma-Aldrich
1-Butene, ≥99%