MilliporeSigma
  • Efficient synthesis of novel NK1 receptor antagonists: selective 1,4-addition of grignard reagents to 6-chloronicotinic acid derivatives.
Efficient synthesis of novel NK1 receptor antagonists: selective 1,4-addition of grignard reagents to 6-chloronicotinic acid derivatives.
The Journal of organic chemistry (2006-02-25)
Fabienne Hoffmann-Emery, Hans Hilpert, Michelangelo Scalone, Pius Waldmeier
ABSTRACT
A new efficient synthesis of two novel classes of NK1 receptor antagonists, among them befetupitant and netupitant, starting from 6-chloronicotinic acid is described. The introduction of the o-tolyl substituent at C4 of the pyridine ring was achieved by a one-pot selective 1,4-Grignard addition/oxidation sequence to 6-chloronicotinic acid or a derivative of it. The scope of this addition/oxidation sequence was examined. It was also shown that the carboxylic function can be converted to a methyl amino group by a Hofmann rearrangement followed by reduction. Furthermore, a new high-yielding synthesis of 2-(3,5-bistrifluoromethylphenyl)-2-methyl propionic acid based on the carbonylation of the tertiary alcohol obtained by Grignard addition of 3,5-bis(trifluoromethyl)bromobenzene to acetone was established.
Sigma-Aldrich
6-Chloropyridine-3-carboxylic acid, 99%