- Identification of the decarboxylated analog of tetrabromotetrachloro-fluorescein and its quantification in the color additives D&C Red Nos. 27 and 28 (phloxine B) using high-performance liquid chromatography.
Identification of the decarboxylated analog of tetrabromotetrachloro-fluorescein and its quantification in the color additives D&C Red Nos. 27 and 28 (phloxine B) using high-performance liquid chromatography.
The present work describes (a) the identification and characterization of an impurity, 2,4,5,7-tetrabromo-6-hydroxy-9-(2,3,4,5-tetrachlorophenyl)-3H-xanthen-3-one (BCPX), in the color additives D&C Red Nos. 27 and 28 (phloxine B) and (b) the determination of the extent and level of BCPX contamination in certified lots of these colors. For these purposes, BCPX (a compound not previously reported in the literature) was synthetically prepared. Test portions from 42 certified lots of D&C Red Nos. 27, 28 and 27 lakes were analyzed for BCPX using an HPLC method that included gradient elution and UV-vis photodiode array detection. Those lots were submitted for certification by both domestic (six) and foreign (six) manufacturers during the past 4 years. Of the test portions analyzed, 32 (76.2%) contained BCPX in amounts ranging from 0.01 to 3.21%. The remaining 10 test portions (23.8%) contained no detectable BCPX or less than 0.008%, which is the limit of quantification for the present method. The analyses revealed substantial differences in the level of BCPX across different manufacturers. The wide range of BCPX levels found in the analyzed lots suggests that the presence of BCPX in D&C Red Nos. 27 and 28 may be avoided or significantly reduced during the manufacturing process.