MilliporeSigma
  • 1,1,1-Tris(hydroxymethyl)propane in manganese carboxylate chemistry: synthesis, structure and magnetic properties of a mixed-valence [MnIII4MnII4] cluster featuring the novel [MnIII4MnII4(mu3-OR)6(mu2-OR)8]6+ core.
1,1,1-Tris(hydroxymethyl)propane in manganese carboxylate chemistry: synthesis, structure and magnetic properties of a mixed-valence [MnIII4MnII4] cluster featuring the novel [MnIII4MnII4(mu3-OR)6(mu2-OR)8]6+ core.
Dalton transactions (Cambridge, England : 2003) (2005-12-21)
Constantinos J Milios, Francesca P A Fabbiani, Simon Parsons, Muralee Murugesu, George Christou, Euan K Brechin
ABSTRACT
The reaction between MnBr(2).4H(2)O with H(3)tmp (1,1,1-tris(hydroxymethyl)propane) in MeCN in the presence of Na(O(2)CCMe(3)) and NBu(4)Br produces the complex [Mn(8)(O(2)CCMe(3))(2)(tmp)(2)(Htmp)(4)Br(4)(H(2)O)(2)].2MeCN (1.2MeCN) in good yield. The centrosymmetric octanuclear molecule consists of four Mn(III) and four Mn(II) ions assembled together by fourteen alkoxo bridges to give a [Mn(III)(4)Mn(II)(4)(mu(3)-OR)(6)(mu(2)-OR)(8)](6+) rod-like core in which the metal centres are arranged in a planar zigzag fashion. Peripheral ligation is provided by a combination of bridging pivalate ions, terminal bromides and water molecules. Dc magnetic susceptibility measurements reveal the presence of dominant antiferromagnetic interactions leading to a spin ground state of S = 0. A rationalization of this result is attempted by structural comparison with previously reported tetranuclear manganese complexes containing the [Mn(III)(2)Mn(II)(2)(mu(3)-OR)(2)(mu(2)-OR)(4)] core in which the magnetic interactions are ferromagnetic.
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, 97%
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, dist., ≥98.0% (GC)