MilliporeSigma
  • The acetates of p-nitrophenyl alpha-L-arabinofuranoside--regioselective preparation by action of lipases.
The acetates of p-nitrophenyl alpha-L-arabinofuranoside--regioselective preparation by action of lipases.
Bioorganic & medicinal chemistry (2005-11-18)
Mária Mastihubová, Jana Szemesová, Peter Biely
ABSTRACT
All possible di-O-acetates and mono-O-acetates of p-nitrophenyl alpha-L-arabinofuranoside were prepared by chemoenzymatic way using lipases. The 2,3-di-O-acetate was obtained in 90% yield by deacetylation of the primary acetyl group of per-O-acetylated p-nitrophenyl alpha-L-arabinofuranoside by Candida cylindracea lipase (CCL) or Candida rugosa lipase (LAY). The 2,5- and 3,5-di-O-acetates were obtained by acetylation of p-nitrophenyl alpha-L-arabinofuranoside by Pseudomonas cepacia lipase (LPS-30) in organic solvents. The 5-O-acetate was regioselectively synthesised in 95% yield by acetylation of p-nitrophenyl alpha-L-arabinofuranoside catalysed by porcine pancreas lipase. Finally, the 2- and 3-O-acetates of p-nitrophenyl alpha-L-arabinofuranoside were obtained in two steps. The enzymatic di-O-acetylation of p-nitrophenyl alpha-L-arabinofuranoside by LPS-30 was followed by enzymatic hydrolysis of the primary acetyl group by CCL or LAY.
Sigma-Aldrich
4-Nitrophenyl α-L-arabinofuranoside, ≥98% (TLC)
Pricing and availability is not currently available.