MilliporeSigma
  • A silychristin isomer and variation of flavonolignan levels in milk thistle (Silybum marianum) fruits.
A silychristin isomer and variation of flavonolignan levels in milk thistle (Silybum marianum) fruits.
Planta medica (2005-10-06)
Wendy A Smith, Denis R Lauren, Elaine J Burgess, Nigel B Perry, Richard J Martin
ABSTRACT
A new natural product has been isolated from silymarin, the hepatoprotective extract of milk thistle (Silybum marianum) fruits. This was shown by NMR spectroscopy to be silychristin B (8), a diastereoisomer of the known silychristin (7). Levels of these and the other flavonolignans varied markedly in fruits of different S. marianum samples.