MilliporeSigma
  • Tryptophan 7-halogenase (PrnA) structure suggests a mechanism for regioselective chlorination.
Tryptophan 7-halogenase (PrnA) structure suggests a mechanism for regioselective chlorination.
Science (New York, N.Y.) (2005-10-01)
Changjiang Dong, Silvana Flecks, Susanne Unversucht, Caroline Haupt, Karl-Heinz van Pée, James H Naismith
ABSTRACT
Chlorinated natural products include vancomycin and cryptophycin A. Their biosynthesis involves regioselective chlorination by flavin-dependent halogenases. We report the structural characterization of tryptophan 7-halogenase (PrnA), which regioselectively chlorinates tryptophan. Tryptophan and flavin adenine dinucleotide (FAD) are separated by a 10 angstrom-long tunnel and bound by distinct enzyme modules. The FAD module is conserved in halogenases and is related to flavin-dependent monooxygenases. On the basis of biochemical studies, crystal structures, and by analogy with monooxygenases, we predict that FADH2 reacts with O2 to make peroxyflavin, which is decomposed by Cl-. The resulting HOCl is guided through the tunnel to tryptophan, where it is activated to participate in electrophilic aromatic substitution.
Sigma-Aldrich
5-Methylindole, 99%