MilliporeSigma
  • Isolation and pharmacological characterization of cannitoxin, a presynaptic neurotoxin from the venom of the Papuan Taipan (Oxyuranus scutellatus canni).
Isolation and pharmacological characterization of cannitoxin, a presynaptic neurotoxin from the venom of the Papuan Taipan (Oxyuranus scutellatus canni).
The Journal of pharmacology and experimental therapeutics (2005-09-02)
Sanjaya Kuruppu, Shane Reeve, Yajnavalka Banerjee, R Manjunatha Kini, A Ian Smith, Wayne C Hodgson
ABSTRACT
The Papuan taipan (Oxyuranus scutellatus canni) is widely distributed throughout much of Papua New Guinea. Although neurotoxicity is a major symptom of envenomation, no neurotoxins have been isolated from this venom. Using a series of size exclusion chromatography steps, we report the isolation of cannitoxin, a presynaptic neurotoxin (44,848 Da) that represents approximately 16% of the whole venom. The toxin displayed high phospholipase A2 (PLA2 activity (330 +/- 5 micromol/min/mg) and caused concentration-dependent (11-66 nM) inhibition of indirect (0.2 ms; 0.1 Hz; supramaximal V) twitches of the chick biventer cervicis nerve-muscle preparation without effecting nicotinic receptor agonists. Prior addition of CSL Taipan antivenom (5 U/ml) or inhibition of phospholipase A2 activity by incubation with 4-bromophenacyl bromide prevented the inhibition of twitches. Cannitoxin is composed of three different subunits, alpha, beta, and gamma, with the possibility of two beta isomers. However, only the alpha subunit displayed in vitro neurotoxic activity of its own. Thus, cannitoxin is similar in structure and pharmacology to taipoxin, which has been isolated from the closely related Australian species O. scutellatus scutellatus (coastal taipan).
Product Description
Supelco
2,4′-Dibromoacetophenone, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%