MilliporeSigma
  • Versatile and stereoselective syntheses of orthogonally protected beta-methylcysteine and beta-methyllanthionine.
Versatile and stereoselective syntheses of orthogonally protected beta-methylcysteine and beta-methyllanthionine.
Organic letters (2005-06-17)
Radha S Narayan, Michael S Vannieuwenhze
ABSTRACT
[reaction: see text] Lantibiotics are a class of lanthionine (and/or beta-methyllanthionine)-containing peptides with antibioitic activity against Gram-positive bacteria. As part of our research effort directed toward the synthesis and mechanistic study of the lantibiotic peptide mersacidin (1), we report stereoselective syntheses of orthogonally protected beta-methylcysteine (beta-MeCys) and beta-methyllanthionine (beta-MeLan), two key nonnatural amino acid components of the mersacidin architecture.
Sigma-Aldrich
S-Methyl-L-cysteine, substrate for methionine sulfoxide reductase A