MilliporeSigma
  • Silicon-directed oxa-Pictet-Spengler cyclization and an unusual dimerization of 2-trimethylsilanyl tryptophols.
Silicon-directed oxa-Pictet-Spengler cyclization and an unusual dimerization of 2-trimethylsilanyl tryptophols.
Organic letters (2005-05-07)
Xuqing Zhang, Xiaojie Li, James C Lanter, Zhihua Sui
ABSTRACT
The tetrahydro-pyrano[3,4-b]indoles 6 were synthesized from 2-(2-trimethylsilanyl-1H-indol-3-yl)-ethanols 5 and various ketones or aldehydes through silicon-directed oxa-Pictet-Spengler cyclizations. An unusual reaction led to the dimeric products 7 when some of 5 was treated with acetone using BF(3) as the catalyst.
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%