MilliporeSigma
  • Evidence for phosphatidylcholine hydrolysis by phospholipase C in rat platelets.
Evidence for phosphatidylcholine hydrolysis by phospholipase C in rat platelets.
Biochimica et biophysica acta (1992-03-25)
E Randell, H Mulye, S Mookerjea, A Nagpurkar
ABSTRACT
Production of [3H]1,2-dipalmitoylglycerol ([3H]DAG) from 1-palmitoyl-2-[9,10-3H]palmitoyl-sn-glycero-3-phosphocholine and [3H]phosphorylcholine from 1,2-dipalmitoyl-sn-glycero-3-[Me-3H]phosphocholine was studied using sonicated rat platelets. The formation of [3H]DAG and [3H]phosphorylcholine occurred at a comparable rate. [3H]Phosphorylcholine formation was dependent on the concentration of the substrate, platelet sonicates and calcium in the incubation medium. The [3H]phosphorylcholine formation increased in presence of 0.01% deoxycholate and 0.01% Triton X-100. The phosphatidylcholine-phospholipase C (PC-PLC) in the platelet sonicates was recovered in both the supernatant and particulate fractions obtained after ultracentrifugation at 105,000 x g for 1 h. The PC-PLC activity in both fractions was inhibited by 2 mM EDTA. In the presence of 0.01% deoxycholate and 0.01% Triton X-100 the activity in the particulate fraction increased compared to the activity in the supernatant, which was inhibited by 0.01% Triton X-100. The pH optima for PC-PLC in both fractions was between pH 7.2 and 7.6. PC-PLC activity was also found in rabbit and human platelet sonicates, but the activity was significantly lower than in rat platelet sonicates. There was no evidence to suggest presence of phosphatidylcholine-specific phospholipase D activity in rat sonicated platelets. This data, therefore, provides direct evidence for the presence of PC-PLC activity in rat platelets.
Sigma-Aldrich
Dipalmitin, ≥99.0%