MilliporeSigma
  • Gas chromatography/mass spectrometry determination of amphetamine-related drugs and ephedrines in plasma, urine and hair samples after derivatization with 2,2,2-trichloroethyl chloroformate.
Gas chromatography/mass spectrometry determination of amphetamine-related drugs and ephedrines in plasma, urine and hair samples after derivatization with 2,2,2-trichloroethyl chloroformate.
Rapid communications in mass spectrometry : RCM (2005-03-05)
Giampietro Frison, Luciano Tedeschi, Donata Favretto, Aikebaier Reheman, Santo Davide Ferrara
ABSTRACT
A new analytical approach, based on derivatization with 2,2,2-trichloroethyl chloroformate and gas chromatography/mass spectrometry (GC/MS), was investigated for qualitative and quantitative analyses of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples. Sample preparation involved alkaline extraction of analytes from biological samples using Extrelut columns, after addition of the internal standard 3,4-methylenedioxypropylamphetamine (MDPA), and subsequent derivatization to produce 2,2,2-trichloroethylcarbamates. GC/MS analyses, in splitless mode using a slightly polar 30-m capillary column, were performed with quadrupole or ion trap instruments. MS acquisition modes were electron ionization (EI) in full-scan or selected ion monitoring (SIM) modes (quadrupole), and full-scan MS or MS/MS modes with chemical ionization (CI) conditions (ion trap). EI spectra of 2,2,2-trichloroethylcarbamates showed variably abundant molecular ions as well as abundant diagnostic fragment ions, both characterized by ion clusters reflecting the isotope distribution of three chlorine atoms in the derivatized molecules. CI spectra showed abundant protonated molecules. Quantitative studies using EI SIM conditions gave recoveries in the range 74-89%, linear response over ranges of 10-2000 ng/mL (plasma and urine) and 0.20-20 ng/mg (hair), with corresponding limits of detection in the ranges 2-5 ng/mL and 0.1-0.2 ng/mg. Potential applications (following full method validation) include clinical and forensic toxicology, as well as doping control.
Sigma-Aldrich
2,2,2-Trichloroethyl chloroformate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2,2,2-Trichloroethyl chloroformate, purum, ≥98.0% (NT)
Pricing and availability is not currently available.