MilliporeSigma
  • Separation of enantiomers in microemulsion electrokinetic chromatography using chiral alcohols as cosurfactants.
Separation of enantiomers in microemulsion electrokinetic chromatography using chiral alcohols as cosurfactants.
Electrophoresis (2004-10-09)
Zhi-Xia Zheng, Jin-Ming Lin, Wing-Hong Chan, Albert W M Lee, Carmen W Huie
ABSTRACT
A novel chiral microemulsion, which involved the use of chiral alcohols as cosurfactants, was demonstrated for the enantiomeric separation of a number of pharmaceutical drugs in microemulsion electrokinetic chromatography (MEEKC). The chiral alcohols investigated were optically active 2-alkanols, with the alkyl chain length having carbon number ranging from 4 to 7. The data indicated that, except for R-(-)-2-butanol, the use of R-(-)-2-pentanol, R-(-)-2-hexanol or R-(-)-2-heptanol as the chiral cosurfactant resulted in the baseline or partial resolution of most of the test solutes, i.e., (+/-)-norephedrine, (+/-)-ephedrine, DL-nadolol, and DL-propranolol. In addition to the chain length of the chiral 2-alkanols, the effects of other experimental conditions, such as the concentration and chirality of the 2-alkanols, as well as the pH of the run buffer and the oil phase of the microemulsion, on the enantiomeric separation of the test solutes were also investigated. An interesting finding was that the water-immiscible organic solvent (oil core) within the microemulsion droplets appeared to play an important role in the chiral separation mechanism. Also, the importance of hydrogen bonding between the test solutes ((+/-)-ephedrine and related compounds) and the chiral microemulsion was demonstrated, as it was not possible to resolve a pair of enantiomers which lacked a beta-amino proton (i.e., (+/-)-N-methyl ephedrine) under optimized run buffer conditions (e.g., 5.0% R-(-)-2-hexanol, 0.8% n-octane, and 3.5% SDS in 90.7% borate buffer at pH 9.2).
Sigma-Aldrich
2-Hexanol, 99%