MilliporeSigma
  • Inhibition of monoamine oxidases by coumarin-3-acyl derivatives: biological activity and computational study.
Inhibition of monoamine oxidases by coumarin-3-acyl derivatives: biological activity and computational study.
Bioorganic & medicinal chemistry letters (2004-06-19)
Franco Chimenti, Daniela Secci, Adriana Bolasco, Paola Chimenti, Arianna Granese, Olivia Befani, Paola Turini, Stefano Alcaro, Francesco Ortuso
ABSTRACT
A series of coumarin-3-acyl derivatives have been synthesized and investigated for the ability to inhibit selectively monoamine oxidases. The coumarin-3-carboxylic acids, 2a-e, proved to be reversible and selective inhibitors of the MAO-B isoform, displaying pIC(50) values of particular interest: 2a shows pIC(50) 7.76 and a selectivity index (pS.I.) 2.94 and 2b shows pIC(50) 7.72 and a pS.I. of 2.80. The coumarin-3-acyl chlorides 3a-e showed high pIC(50) values against both MAO-A and MAO-B isoforms, 3d being the highest against MAO-B with a pIC(50) value of 8.00. In order to rationalize the activity/selectivity results, molecular descriptors were generated. Further insight about enzyme-inhibitor interaction was obtained by docking experiments with the MAO-B isoform.
Sigma-Aldrich
Coumarin-3-carboxylic acid, 99%
Pricing and availability is not currently available.