MilliporeSigma
  • Synthesis, self-assembly and characterization of a new glucoside-type hydrogel having a Schiff base on the aglycon.
Synthesis, self-assembly and characterization of a new glucoside-type hydrogel having a Schiff base on the aglycon.
Carbohydrate research (2004-04-29)
Chunyan Bao, Ran Lu, Ming Jin, Pengchong Xue, Changhui Tan, Yingying Zhao, Guofa Liu
ABSTRACT
A new hydrogel based on a substituted phenyl glucoside with a Schiff base in the aglycon was synthesized, and the self-assembling characteristics was studied. FTIR spectra, UV-vis absorption spectra and X-ray diffraction (XRD) revealed that pi-pi interactions between the Schiff base moieties, hydrogen bonds, and the interdigitated interactions between hydrophobic chains had effects on the formation of the self-assembling hydrogel. Scanning electron microscopic (SEM) and transmission electron microscopic (TEM) observation showed that the three-dimensional hydrogel network was constructed from nanotubes with inner diameters of ca. 75 nm and wall of ca. 20 nm.
Sigma-Aldrich
Phenyl β-D-glucopyranoside, ≥95.0%
Pricing and availability is not currently available.