MilliporeSigma
Exposure to Asulox inhibits the growth of mosses.
Annals of botany (2003-08-23)
J K Rowntree, K F Lawton, F J Rumsey, E Sheffield
ABSTRACT
Asulox is a herbicide used to control bracken. Its effects on mosses were investigated to ascertain whether exposure proved as detrimental as found in parallel studies on pteridophytes. Mature gametophytes of 18 mosses were exposed to a range of concentrations of Asulox under standard conditions and the effects on growth monitored. Plants were cut to a standard length, exposed to Asulox solution for 24 h, grown for 3 weeks and total elongation (main stem and branches) measured. EC50 values were calculated and species ranked according to sensitivity. The effects of exposure on total elongation were compared with those on main stem elongation alone. Under the conditions tested, the total elongation of all species was inhibited after exposure to Asulox. The amount of elongation observed after exposure was different for different species and inhibition of elongation occurred at different exposure concentrations. A single regression equation was not adequate to describe the dose response curves of all species tested. An ability to produce secondary branches may confer increased tolerance to Asulox exposure. It is concluded that mosses suffer detrimental effects after exposure to Asulox at concentrations similar to those that affect fern gametophytes such as bracken.