MilliporeSigma
  • [Fluorescence enhancement character of terbium perchlorate and thulium perchlorate with phenylcarboxymethyl sulfoxide coordination compounds].
[Fluorescence enhancement character of terbium perchlorate and thulium perchlorate with phenylcarboxymethyl sulfoxide coordination compounds].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-14)
Wen-xian Li, Guo-jiun Wu, Zhong-shi Liu, Feng-mei Han
ABSTRACT
(Tb1-x Tmx).L2.(ClO4).2H2O(x = 0.000 to 0.200, L = C6H5SOCH2COO-) have been synthesized. The coordination compounds have been studied by means of composition analysis, molar conductivity, IR, and the condition of coordination have been inferred. In the fluorescent spectra it was found that Tm3+ has a strongly sensitization effect to the fluorescence of Tb3+. The fluorescent emission intensity of Terbium perchlorate with phenylcarboxymethyl sulfoxide coordination compounds would be enhanced by Tm3+ in mixing. Tm3+ has a sensitization to the fluorescence of Tb3+ in the ratio of Tb3+:Tm3+ = 0.999:0.001-0.900:0.100. In the solubility experiment it was found that the complexes have high solubility in ethanol.
Sigma-Aldrich
Diphenyl sulfoxide, 96%