MilliporeSigma
  • Enantiomeric differentiation of bornyl acetate by 13C-NMR using a chiral lanthanide shift reagent.
Enantiomeric differentiation of bornyl acetate by 13C-NMR using a chiral lanthanide shift reagent.
Phytochemical analysis : PCA (2003-08-02)
Nicolas Baldovini, Félix Tomi, Joseph Casanova
ABSTRACT
The enantiomeric differentiation of bornyl acetate was carried out by 13C-NMR spectroscopy using a chiral lanthanide shift reagent. The technique was successfully applied to the determination of the enantiomer of bornyl acetate present in the essential oil of Inula graveolens.