MilliporeSigma
  • Synthesis and evaluation of a C8 stationary phase on a silica hydride surface by hydrosilation of 1-octyne.
Synthesis and evaluation of a C8 stationary phase on a silica hydride surface by hydrosilation of 1-octyne.
Journal of chromatography. A (2003-05-09)
Joseph J Pesek, Maria T Matyska, Xiaofan Pan
ABSTRACT
The silanization/hydrosilation method is used to bond an alkyne (1-octyne) to a silica hydride surface. The new bonded material is characterized by elemental analysis and diffuse reflectance infrared Fourier transform spectroscopy. The hydrophobic behavior of this material was determined by the retention characteristics of aromatic solutes and the shape selectivity as well as phase classification (monomeric or polymeric) was measured by the polycyclic aromatic hydrocarbon mixture standard reference material 869. The presence of residual silanols on the bonded phase was probed by several basic solutes at pH 7. Long-term stability studies were conducted by measuring retention and peak symmetry of basic compounds over several thousand column volumes at pH 10.
Sigma-Aldrich
1-Octyne, 97%
Pricing and availability is not currently available.