MilliporeSigma
  • Isozyme-specific induction of low-dose aspirin on cytochrome P450 in healthy subjects.
Isozyme-specific induction of low-dose aspirin on cytochrome P450 in healthy subjects.
Clinical pharmacology and therapeutics (2003-03-07)
Xiao-Ping Chen, Zhi-Rong Tan, Song-Lin Huang, Zheng Huang, Dong-Sheng Ou-Yang, Hong-Hao Zhou
ABSTRACT
This study was designed to define the effect of low-dose aspirin administration on the activity of cytochrome P450 (CYP) in normal human subjects. Aspirin, 50 mg daily, was given for 14 days to 18 nonsmoking healthy male volunteers. A modified 5-drug cocktail procedure consisting of caffeine, mephenytoin, metoprolol, chlorzoxazone, and midazolam was performed to simultaneously assess in vivo activity of CYP1A2, CYP2C19, CYP2D6, CYP2E1, and CYP3A, respectively. The activities were assessed on 4 occasions including at baseline, after 7 and 14 daily doses of aspirin, and at 7 days after discontinuation of aspirin. Concentrations of parent drugs and corresponding metabolites in biologic samples were assayed by reversed-phase HPLC. Both 7-day and 14-day aspirin intake increased the activity of CYP2C19 significantly, as indicated by 4-hydroxymephenytoin urinary recovery (P <.001). Induction of low-dose aspirin on CYP2C19 was time-dependent. CYP3A activity indices increased moderately but significantly by both 7-day and 14-day aspirin treatment (P <.05), but the percentage changes in CYP3A activity indices were not significant. Low-dose aspirin had no effect on CYP1A2, CYP2D6, and CYP2E1 in vivo activity by either 7-day or 14-day treatment. The effect of low-dose aspirin on CYPs was enzyme-specific. Both 7-day and 14-day low-dose aspirin induced the in vivo activities of CYP2C19 but did not affect the activities of CYP1A2, CYP2D6, and CYP2E1. The effect of low-dose aspirin on CYP3A activity awaits further confirmation. When low-dose aspirin is used in combination with drugs that are substrates of CYP2C19, doses of the latter should be adjusted to ensure their efficacy.
Sigma-Aldrich
4-Hydroxymephenytoin
Sigma-Aldrich
4-Hydroxymephenytoin, ≥98% (HPLC)