MilliporeSigma
  • rH-metric controls and primary standardization in aqueous-organic media.
rH-metric controls and primary standardization in aqueous-organic media.
Annali di chimica (2002-12-20)
Luigi Falciola, Patrizia R Mussini, Torquato Mussini
ABSTRACT
The rationale for the extension of the rH-metric standardization and measurements to aqueous-organic solvent mixtures is here introduced, and examples of establishment of the ranges of conventional rH-metric scales as well as primary standards rHs in some typical aqueous-organic solvent mixtures are described.
Sigma-Aldrich
Quinhydrone, 97%