MilliporeSigma
  • A transcriptional luxAB reporter fusion responding to fluorene in Sphingomonas sp. LB126 and its initial characterisation for whole-cell bioreporter purposes.
A transcriptional luxAB reporter fusion responding to fluorene in Sphingomonas sp. LB126 and its initial characterisation for whole-cell bioreporter purposes.
Research in microbiology (2002-01-05)
L Bastiaens, D Springael, W Dejonghe, P Wattiau, H Verachtert, L Diels
ABSTRACT
The promoter probe mini-Tn5-luxAB-tet was used to create a luxAB transcriptional fusion responding to fluorene in the fluorene utilising bacterium Sphingomonas sp. LB126. The mutant strain, named L-132, was impaired in fluorene utilisation and strongly emitted light upon addition of fluorene to the growth medium. L-132 was initially characterised and examined for its potential use as a whole-cell biosensor in the perspective of quantifying fluorene in environmental samples. Activity of the reporter gene as a response to fluorene was detectable after 30 min and was optimal after 4 h. A linear response to fluorene concentrations within the water solubility range was achieved, with a detection limit of 200 microg per litre. Besides fluorene, L-132 weakly responded to the polycyclic aromatic hydrocarbons phenanthrene and dibenzothiophene, whereas strong responses were obtained with 9-fluorenone, 9-hydroxyfluorene, phthalic acid and protocatechuic acid. The latter four compounds are metabolites formed in course of fluorene degradation, which suggested that a fluorene metabolite rather than fluorene itself was the true inducer of the luxAB fusion in L-132.
Sigma-Aldrich
9-Hydroxyfluorene, 96%
Pricing and availability is not currently available.